missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-12 (p70) ELISA Kits

Product Code. 10465725
Click to view available options
Quantity:
1 Plate Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10465725

Brand: Invitrogen™ EHIL12

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Measure levels of active human interleukin-12, p70 in human serum, plasma, urine and culture supernatant samples using 96-well plates.

Assay IL-12 p70 (active human interleukin-12, p70) levels in human serum, plasma (EDTA, heparin, sodium citrate), urine and culture supernatant samples with our Thermo Scientific Human IL-12 (p70) ELISA Kits.

These human IL-12 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of biologically active interleukin-12 (IL-12 p70) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Biologically active (p70) natural and recombinant human interleukin-12 (IL-12, p70)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 15.4 to 600pg/mL

  • Sensitivity: < 3pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with human IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-11, IL-13, IL-15, TNFα, IFNγ, GM-CSF, GROα or GROβ. Recombinant human IL-12 (p40) and mouse IL-12 (p40) do not cross-react or interfere with this assay

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.4 to 600pg/mL
Assay Sensitivity <3pg/mL
Description Human IL-12 (p70) ELISA Kit
Immunoassay Kit Format Colorimetric ELISA (TMB substrate) with Plates
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine
Specificity No significant cross-reaction with human IL-1a, IL-1ß, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-11, IL-13, IL-15, TNFa, IFNg, GM-CSF, GROa or GROß. Recombinant human IL-12 (p40) and mouse IL-12 (p40) do not cross-react or interfere with t
Sufficient For 96 microplate wells
Kit Contents 30X Wash Buffer, 50mL, Stop Solution, 13mL, Adhesive Plate Covers, 6 ea, Biotinylated Antibody Reagent, 8mL, HRP Dilution Buffer, 13mL, Anti-Human IL-12 Precoated 96-well Strip Plate, 1 ea, Standard Diluent, 14mL, Streptavidin-HRP Concentrate, 75 μL, TMB Substrate Solution, 13mL, Lyophilized Recombinant Human IL-12 Standard, 2 vials
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Intra-assay CV <10%; inter-assay CV <10%
Quantity 1 Plate Kit
Target Biologically active (p70) natural and recombinant human interleukin-12 (IL-12, p70)
Total Assay Time 3 hr.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.