Learn More
R&D Systems™ Human IL-10 XL Luminex™ Performance Assay
Product Code. 30001342
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001342 1 Kit 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30001342 Supplier R&D Systems™ Supplier No. LUXLM217
Specifications
Product Type Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Quantity 1 Kit
Species Human
Binding Target IL-10
Gene ID (Entrez) 3586
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.