Learn More
R&D Systems™ Human IL-10 Quantikine ELISA Kit
Product Code. 18792294
Change view
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 Plate
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18792294 1 Pk. (6 Plates) 1 set
18752064 1 Plate 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18792294 Supplier R&D Systems™ Supplier No. S1000B
  • ONLINE EXCLUSIVE PRICE
    4585.00€
    4333.35€ / 1 sets
    Save 251.65€
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 28-09-2026
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Human IL-10 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Human IL-10 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-10 in cell culture supernates (200 ul), serum (200 ul), edta plasma (200 ul), heparin plasma (200 ul), citrate plasma (200 ul).
TRUSTED_SUSTAINABILITY
Specifications
Assay Range 7.80 to 500 pg/mL
Assay Sensitivity 3.9 pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Line R&D Systems
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Citrate, Plasma (EDTA, Citrate, Heparin), Serum
Specificity Natural and Recombinant Human IL-10
Sufficient For 6 x 96-well Plates
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3586
Gene Symbol IL10
Quantity 1 Pk. (6 Plates)
Sample Volume Cell Culture Supernates (200uL),Serum (200uL),EDTA Plasma (200uL),Heparin Plasma (200uL),Citrate Plasma (200uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10 mgC126451, interleukin 10, interleukin-10, TGIF
Target IL-10