Learn More
R&D Systems™ Human IL-10 High Sensitivity Magnetic Luminex™ Performance Assay
Product Code. 18461810
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18461810 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18461810 Supplier R&D Systems™ Supplier No. LHSCM217
  • ONLINE EXCLUSIVE PRICE
    261.00€
    246.75€ / 1 sets
    Save 14.25€
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
  • Magnetic format allows for easier wash steps
  • Undergoes similar validation testing as Quantikine™ ELISA Kits
  • Precise and Specific
  • Requires a small sample volume (<50 μL)
  • Run your assay in just 3-5 hours
  • Mass-calibrated standards for consistent results with every new lot of material
  • Enough reagents for one 96-well plate
TRUSTED_SUSTAINABILITY
Specifications
Product Type Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Quantity 1 Kit
Species Human
Binding Target IL-10
Bead Type Magnetic
Assay Sensitivity 0.24pg/mL
Assay Range 0.46 - 1900pg/mL
Gene ID (Entrez) 3586
Gene Symbol IL10
Sample Type Serum, Plasma (EDTA, Heparin)
Sample Volume Serum (50μL),EDTA Plasma (50μL),Heparin Plasma (50μL)
Synonym CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Kit Contents
  • Microparticle Concentrate
  • Biotin-Antibody Concentrate
Content And Storage Store the unopened product at 2° to 8°C. Do not use past expiration date.