missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human IL-10 Detection Kit (Flow Cytometry Bead Assay)
Description
Human IL-10 Detection Kit (Flow Cytometry Bead Assay) is expressed from Human
Specifications
Specifications
| Quantity | 100 Tests |
| Assay Sensitivity | 0.1 pg/mL |
| Sample Type | Human Serum, Human Plasma, Cell Culture Supernatants |
| Sample Volume | 30 μL |
| Content And Storage | 1. Unopened buffer should be stored at 2°C to 8°C upon receiving. 2. Find the expiration date on the outside packaging and do not use reagents past their expiration date. 3. The shelf life is 30 days from the date of opening. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?