Learn More
R&D Systems™ Human IGSF8/CD316 (NP_443100) VersaClone cDNA
Product Code. 18713197
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18713197 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18713197 Supplier R&D Systems™ Supplier No. RDC1241
Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.
  • The Human IGSF8/CD316 (NP_443100) VersaClone cDNA clone contains the complete ORF for IGSF8/CD316, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The IGSF8/CD316 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY
Specifications
Gene hIGSF8/CD316
Accession Number NP_443100
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gene ID (Entrez) 93185
Content And Storage Store the unopened product at -20 to -80°C
Species Human
Quantity 10 μg