Learn More
Novus Biologicals™ Human IgM Native Protein
Product Code. 18445237 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 18445237
Brand: Novus Biologicals™ NBP1970561mg
Specifications
For Use With (Application) Western Blot, ELISA, Immunohistochemistry, SDS-PAGE
Formulation 0.1 M Tris Chloride, 0.5 M Sodium Chloride, pH 8.0
Quantity 1 mg
Storage Requirements Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze/thaw cycles.
Gene Alias AGM1, MU, VH
Inhibitor Properties This product has been tested in SDS-Page and can be used as a control or standard in indirect trapping ELISA for quantitation of antigen in serum using a standard curve, for immunoprecipitation and for western blotting.
Purity or Quality Grade Multi-step
Protein Human IgM