Learn More
STEMCELL Technologies Human IgE ELISA Kit
Product Code. 17158041
Encompass_Preferred
Click to view available options
Quantity:
2 Plates
Unit Size:
Pack of 2
Product Code. 17158041
Brand: STEMCELL Technologies 02032
For detection and measurement of human immunoglobulin E
  • The Human Immunoglobulin E (IgE) ELISA Kit is designed for the quantitative detection and measurement of human IgE in biological fluids such as serum, plasma, and cell culture supernatants
  • IgE is found in the lungs, skin, and mucous membranes
  • IgE plays a role in allergic reactions by stimulating degranulation of eosinophils, mast cells, and basophils through interaction with the Fc epsilon Receptor I (FcεRI)
  • The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the immunoglobulin
  • The captured immunoglobulin is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase (SA-HRP), which binds the biotinylated antibody
  • Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of immunoglobulin in the sample
Specifications
Description B Cells; Hybridomas
Quantity 2 Plates