Learn More
STEMCELL Technologies Human IgE ELISA Kit
Click to view available options
Quantity:
2 Plates
Unit Size:
Pack of 2
Description
- The Human Immunoglobulin E (IgE) ELISA Kit is designed for the quantitative detection and measurement of human IgE in biological fluids such as serum, plasma, and cell culture supernatants
- IgE is found in the lungs, skin, and mucous membranes
- IgE plays a role in allergic reactions by stimulating degranulation of eosinophils, mast cells, and basophils through interaction with the Fc epsilon Receptor I (FcεRI)
- The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the immunoglobulin
- The captured immunoglobulin is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase (SA-HRP), which binds the biotinylated antibody
- Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of immunoglobulin in the sample
Specifications
Specifications
| Description | B Cells; Hybridomas |
| Quantity | 2 Plates |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.