missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human IFT122 Partial ORF (NP_443711, 1194 a.a. - 1291 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055764-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16158356

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This cytoplasmic protein contains seven WD repeats and an AF-2 domain which function by recruiting coregulatory molecules and in transcriptional activation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]

Sequence: SIGDEDPFTAKLSFEQGGSEFVPVVVSRLVLRSMSRRDVLIKRWPPPLRWQYFRSLLPDASITMCPSCFQMFHSEDYELLVLQHGCCPYCRRCKDDPG
Specifications

Specifications

NP_443711
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SIGDEDPFTAKLSFEQGGSEFVPVVVSRLVLRSMSRRDVLIKRWPPPLRWQYFRSLLPDASITMCPSCFQMFHSEDYELLVLQHGCCPYCRRCKDDPG
RUO
IFT122
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
55764
IFT122 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SPG/WDR10/WDR10p/WDR140
IFT122
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers