Learn More
Abnova™ Human IFT122 Partial ORF (NP_443711, 1194 a.a. - 1291 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055764-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This cytoplasmic protein contains seven WD repeats and an AF-2 domain which function by recruiting coregulatory molecules and in transcriptional activation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: SIGDEDPFTAKLSFEQGGSEFVPVVVSRLVLRSMSRRDVLIKRWPPPLRWQYFRSLLPDASITMCPSCFQMFHSEDYELLVLQHGCCPYCRRCKDDPGSpecifications
NP_443711 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SIGDEDPFTAKLSFEQGGSEFVPVVVSRLVLRSMSRRDVLIKRWPPPLRWQYFRSLLPDASITMCPSCFQMFHSEDYELLVLQHGCCPYCRRCKDDPG | |
RUO | |
IFT122 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55764 | |
IFT122 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SPG/WDR10/WDR10p/WDR140 | |
IFT122 | |
Recombinant | |
wheat germ expression system |