missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IFN-beta Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 96
Description
The Human IFN-beta Quantikine ELISA Kit quantifies human IFN-beta in cell culture supernates (50 ul).
Specifications
Specifications
| Assay Range | 7.81 - 500pg/mL |
| Assay Sensitivity | 0.781pg/mL |
| Sample Type | Cell Culture Supernatant |
| Specificity | Natural and Recombinant Human IFN-beta |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3456 |
| Quantity | 1 Kit |
| Sample Volume | Cell Culture Supernates (50uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction