missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human I-TAC ELISA Kits

Product Code. 12341873
Click to view available options
Quantity:
1 Plate Kit
Unit Size:
Each
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 12341873

Brand: Invitrogen™ EHITAC

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Measure levels of human interferon-inducible T-cell alpha chemoattractant in human serum, plasma and culture supernatant samples using 96-well plates.

Assay I-TAC (human interferon-inducible T-cell alpha chemoattractant) levels in human serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples with our Invitrgen™ Human I-TAC ELISA Kits.

These human I-TAC ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of I-TAC (also known as CXCL11 or IP9) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant human I-TAC (human interferon-inducible T-cell alpha chemoattractant, also called CXCL11, IP9)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 41.0 to 4000pg/mL

  • Sensitivity: < 5pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3.5 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with human MIG/CXCL9, IP-10/CXCL10, IL-8, IFNα or mouse ITAC/CXCL11

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 41.0 to 4000pg/mL
Assay Sensitivity <5pg/mL
Description Human I-TAC ELISA Kit
Immunoassay Kit Format Colorimetric ELISA (TMB substrate) with Plates
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificity No significant cross-reaction with human MIG/CXCL9, IP-10/CXCL10, IL-8, IFNa or mouse ITAC/CXCL11
Sufficient For 96 microplate wells
Kit Contents Streptavidin-HRP Concentrate, 75 μL, TMB Substrate Solution, 13mL, Stop Solution, 13mL, Standard Diluent, 28mL, 30X Wash Buffer, 50mL, Adhesive Plate Covers, 6 ea, Anti-Human I-TAC Precoated 96-well Strip Plate, 1 ea, Biotinylated Antibody Reagent, 8mL, Lyophilized Recombinant Human I-TAC Standard, 2 vials, HRP Dilution Buffer, 14mL
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Intra-assay CV <10%; inter-assay CV <10%
Quantity 1 Plate Kit
Sample Volume 50 μL
Target Natural and recombinant human I-TAC (human interferon-inducible T-cell alpha chemoattractant, also called CXCL11, IP9)
Total Assay Time 3.5 hr.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.