missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human HVEM/TNFRSF14 DuoSet ELISA

Product Code. 30021147
Click to view available options
Quantity:
1 Kit (for 15 Plates)
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30021147

missing translation for 'mfr': R&D Systems™ DY356

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sufficient for at least 15 96-well plates

The Human HVEM/TNFRSF14 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human HVEM/TNFRSF14.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit (for 15 Plates)
Species Human
Conjugate Biotin
Assay Range 15.60 to 1000 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 8764
Synonym ATAR, CD270, CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, TNFRSF14, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.