Learn More
Abnova™ Human HTR2B Partial ORF (AAH63123.1, 199 a.a. - 359 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003357-Q02.10ug
Additional Details : Weight : 0.02000kg
Description
Multiple receptor subtypes of serotonin neurotransmitters with multiple physiologic functions have been recognized. The 5-HT-2 receptors mediate many of the central and peripheral physiologic functions of serotonin. Cardiovascular effects include contraction of blood vessels and shape changes in platelets; central nervous system effects include neuronal sensitization to tactile stimuli and mediation of hallucinogenic effects of phenylisopropylamine hallucinogens.[supplied by OMIM]
Sequence: VDNPNNITCVLTKERFGDFMLFGSLAAFFTPLAIMIVTYFLTIHALQKKAYLVKNKPPQRLTWLTVSTVFQRDETPCSSPEKVAMLDGSRKDKALPNSGDETLMRRTSTIGKKSVQTISNEQRASKVLGIVFFLFLLMWCPFFITNITLVLCDSCNQTTLQSpecifications
AAH63123.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
43.45kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VDNPNNITCVLTKERFGDFMLFGSLAAFFTPLAIMIVTYFLTIHALQKKAYLVKNKPPQRLTWLTVSTVFQRDETPCSSPEKVAMLDGSRKDKALPNSGDETLMRRTSTIGKKSVQTISNEQRASKVLGIVFFLFLLMWCPFFITNITLVLCDSCNQTTLQ | |
RUO | |
HTR2B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3357 | |
HTR2B (Human) Recombinant Protein (Q02) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
5-HT(2B)/5-HT2B | |
HTR2B | |
Recombinant | |
wheat germ expression system |