missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HTR1A Partial ORF (NP_000515, 1 a.a. - 36 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003350-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MDVLSPGQGNNTTSPPAPFETGGNTTGISDVTVSYQSpecifications
NP_000515 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
29.7kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDVLSPGQGNNTTSPPAPFETGGNTTGISDVTVSYQ | |
RUO | |
HTR1A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3350 | |
HTR1A (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
5-HT1A/5HT1a/ADRB2RL1/ADRBRL1 | |
HTR1A | |
Recombinant | |
wheat germ expression system |