Learn More
Abcam Human HSP90AB1 (Hsp90 beta) knockout A-431 cell line
Product Code. 32154108
Change view
Click to view available options
Quantity:
1,000,000 Cells
2 x 1,000,000 Cells
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32154108 2 x 1,000,000 Cells Pack of 2
32139315 1,000,000 Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 32154108 Supplier Abcam Supplier No. ab2694912x1000000Cells_
 Restricted Item
Estimated Shipment: 06-10-2026
HSP90AB1 KO cell line available to order. KO validated by Next Generation Sequencing, Western blot. Free of charge wild type control available. Knockout achieved by CRISPR/Cas9; X = 8 bp deletion, 22 bp deletion; Frameshift: 100%. To order both knockout and wild-type control cells: select 2 x 10...
HSP90AB1 KO cell line available to order. KO validated by Next Generation Sequencing, Western blot. Free of charge wild type control available. Knockout achieved by CRISPR/Cas9; X = 8 bp deletion, 22 bp deletion; Frameshift: 100%. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells