Learn More
ACROBiosystems Human HSP90AA1 / HSP86 Protein, GST Tag
Brand:  ACROBiosystems HS1-H513X-100UG
Additional Details : Weight : 0.00001kg
 View more versions of this product
Product Code. 15806947
  • 326.60€ / 100µg
Estimated Shipment: 09-05-2024
to see stock.
Add to basket
Description
Description
Heat shock protein HSP 90-alpha (HSP90AA1 or HSP90A) is also known as Heat shock 86 kDa (HSP 86 or HSP86), Renal carcinoma antigen NY-REN-38, HSPC1, HSPCA, EL52, HSP89A, HSP90N, HSPCAL1, HSPCAL4, HSPN, Hsp89, Hsp90, LAP2, which belongs to the heat shock protein 90 family. HSP90AA1 undergoes a functional cycle that is linked to its ATPase activity. This cycle probably induces conformational changes in the client proteins, thereby causing their activation. HSP90AA1 interacts dynamically with various co-chaperones that modulate its substrate recognition, ATPase cycle and chaperone function.
This protein carries a GST tag at the N-terminus. The protein has a calculated MW of 49.3 kDa. The protein migrates as 49 kDa under reducing (R) condition (SDS-PAGE).
Specifications
Specifications
NP_005339
100 μg
Glu 535 - Asp 732
HSP90AA1,HSP90A,HSP 86,HSP86,HSPC1,HSPCA,EL52,HSP89A,HSP90N,HSPCAL1,HSPCAL4,HSPN,Hsp89,Hsp90,LAP2
Unconjugated
GST Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>85% as determined by SDS-PAGE
49.3 kDa
Human HSP90AA1, GST Tag (HS1-H513x) is expressed from E.coli cells. It contains AA Glu 535 - Asp 732 (Accession # NP_005339).
-20°C
HSP90AA1
Human
50 mM HEPES, 150 mM NaCl, pH7.0
E.coli
Bio-Markers and CD Antigens