Learn More
Abnova™ Human HSD17B3 Partial ORF (NP_000188, 29 a.a. - 119 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003293-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This isoform of 17 beta-hydroxysteroid dehydrogenase is expressed predominantly in the testis and catalyzes the conversion of androstenedione to testosterone. It preferentially uses NADP as cofactor. Deficiency can result in male pseudohermaphroditism with gynecomastia. [provided by RefSeq]
Sequence: RCVLLNYWKVLPKSFLRSMGQWAVITGAGDGIGKAYSFELAKRGLNVVLISRTLEKLEAIATEIERTTGRSVKIIQADFTKDDIYEHIKEKSpecifications
NP_000188 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.75kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RCVLLNYWKVLPKSFLRSMGQWAVITGAGDGIGKAYSFELAKRGLNVVLISRTLEKLEAIATEIERTTGRSVKIIQADFTKDDIYEHIKEK | |
RUO | |
HSD17B3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3293 | |
HSD17B3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EDH17B3/SDR12C2 | |
HSD17B3 | |
Recombinant | |
wheat germ expression system |