missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HOXD4 Partial ORF (NP_055436.2, 1 a.a. - 62 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003233-Q01.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16094175

  • 508.00€ / 25µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located at 2q31-2q37 chromosome regions. Deletions that removed the entire HOXD gene cluster or 5' end of this cluster have been associated with severe limb and genital abnormalities. The protein encoded by this gene may play a role in determining positional values in developing limb buds. Alternatively spliced variants have been described but their full length nature has not been determined. [provided by RefSeq]

Sequence: MVMSSYMVNSKYVDPKFPPCEEYLQGGYLGEQGADYYGGGAQGADFQPPGLYPRPDFGEQPF
Specifications

Specifications

NP_055436.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.56kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MVMSSYMVNSKYVDPKFPPCEEYLQGGYLGEQGADYYGGGAQGADFQPPGLYPRPDFGEQPF
RUO
HOXD4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3233
HOXD4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HHO.C13/HOX-5.1/HOX4/HOX4B/Hox-4.2
HOXD4
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers