missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HOXC6 Full-length ORF (NP_004494.1, 1 a.a. - 235 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003223-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16196321

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene belongs to the homeobox family, members of which encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene, HOXC6, is one of several HOXC genes located in a cluster on chromosome 12. Three genes, HOXC5, HOXC4 and HOXC6, share a 5' non-coding exon. Transcripts may include the shared exon spliced to the gene-specific exons, or they may include only the gene-specific exons. Alternatively spliced transcript variants encoding different isoforms have been identified for HOXC6. Transcript variant two includes the shared exon, and transcript variant one includes only gene-specific exons. [provided by RefSeq]

Sequence: MNSYFTNPSLSCHLAGGQDVLPNVALNSTAYDPVRHFSTYGAAVAQNRIYSTPFYSPQENVVFSSSRGPYDYGSNSFYQEKDMLSNCRQNTLGHNTQTSIAQDFSSEQGRTAPQDQKASIQIYPWMQRMNSHSGVGYGADRRRGRQIYSRYQTLELEKEFHFNRYLTRRRRIEIANALCLTERQIKIWFQNRRMKWKKESNLTSTLSGGGGGATADSLGGKEEKREETEEEKQKE
Specifications

Specifications

NP_004494.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
53.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CP25/HHO.C8/HOX3/HOX3C
HOXC6
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
3223
HOXC6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNSYFTNPSLSCHLAGGQDVLPNVALNSTAYDPVRHFSTYGAAVAQNRIYSTPFYSPQENVVFSSSRGPYDYGSNSFYQEKDMLSNCRQNTLGHNTQTSIAQDFSSEQGRTAPQDQKASIQIYPWMQRMNSHSGVGYGADRRRGRQIYSRYQTLELEKEFHFNRYLTRRRRIEIANALCLTERQIKIWFQNRRMKWKKESNLTSTLSGGGGGATADSLGGKEEKREETEEEKQKE
RUO
HOXC6
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers