missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HKR2 Partial ORF (NP_862829.1, 196 a.a. - 294 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00342945-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192837

  • 528.00€ / 25µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

Sequence: KSVTQQIHFKKTSGPYKDVPTDQRGRESGASRNSSSAWPNLTSQEKPPSEDKFDLVDAYGTEPPYTYSGKRSSKCRECRKMFQSASALEAHQKTHSRKT
Specifications

Specifications

NP_862829.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KSVTQQIHFKKTSGPYKDVPTDQRGRESGASRNSSSAWPNLTSQEKPPSEDKFDLVDAYGTEPPYTYSGKRSSKCRECRKMFQSASALEAHQKTHSRKT
RUO
ZSCAN22
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
342945
HKR2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HKR2/MGC126679/MGC138482/ZNF50
ZSCAN22
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers