Learn More
R&D Systems™ Human HFE (NP_000401.1) VersaClone cDNA
Specifications
Specifications
| Gene | HFE |
| Accession Number | NP_000401.1 |
| Concentration | 10 μg (at 0.2 μg/μL) |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1060 bp |
| Gene ID (Entrez) | 3077 |
| Gene Alias | dJ221C16.10.1, hemochromatosis, hereditary hemochromatosis protein, hereditary hemochromatosis protein HLA-H, HFE1, HH, high Fe, HLAH, HLA-H, HLA-HHFE1, MGC103790, MHC class I-like protein HFE, MVCD7, TFQTL2 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |