Learn More
Abnova™ Human HBXAP Partial ORF (NP_057662.3, 1343 a.a. - 1441 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051773-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
HBXAP is involved in transcription repression, transcription coactivation when associated with hepatitis B virus X protein (HBX), and chromatin remodeling and spacing when associated with SNF2H (MIM 603375).[supplied by OMIM]
Sequence: IESDEEEDFENVGKVGSPLDYSLVDLPSTNGQSPGKAIENLIGKPTEKSQTPKDNSTASASLASNGTSGGQEAGAPEEEEDELLRVTDLVDYVCNSEQLSpecifications
NP_057662.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IESDEEEDFENVGKVGSPLDYSLVDLPSTNGQSPGKAIENLIGKPTEKSQTPKDNSTASASLASNGTSGGQEAGAPEEEEDELLRVTDLVDYVCNSEQL | |
RUO | |
RSF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51773 | |
HBXAP (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HBXAP/RSF-1/XAP8/p325 | |
RSF1 | |
Recombinant | |
wheat germ expression system |