missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HARSL Partial ORF (NP_036340, 2 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_036340 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23438 |
Molecular Weight (g/mol) | 37.73kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16154686
|
Abnova™
H00023438-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
16164686
|
Abnova™
H00023438-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
Description
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is an enzyme belonging to the class II family of aminoacyl-tRNA synthetases. Functioning in the synthesis of histidyl-transfer RNA, the enzyme plays an accessory role in the regulation of protein biosynthesis. The gene is located in a head-to-head orientation with HARS on chromosome five, where the homologous genes share a bidirectional promoter. [provided by RefSeq]
Sequence: PLLGLLPRRAWASLLSQLLRPPCASCTGAVRCQSQVAEAVLTSQLKAHQEKPNFIIKTPKGTRDLSPQHMVVREKILDLVISCFKRHGAKGMDTPAFELKETLTEKYGESpecifications
NP_036340 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HARSL/HARSR/HO3 | |
HARS2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
23438 | |
HARSL (Human) Recombinant Protein (Q01) | |
PLLGLLPRRAWASLLSQLLRPPCASCTGAVRCQSQVAEAVLTSQLKAHQEKPNFIIKTPKGTRDLSPQHMVVREKILDLVISCFKRHGAKGMDTPAFELKETLTEKYGE | |
RUO | |
HARS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |