missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HAPLN4 Partial ORF (NP_075378, 30 a.a. - 134 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_075378 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 404037 |
Molecular Weight (g/mol) | 37.29kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16112987
|
Abnova™
H00404037-Q01.25ug |
25 ug |
508.00€
25µg |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
16102987
|
Abnova™
H00404037-Q01.10ug |
10 ug |
335.00€
10µg |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
Description
Sequence: QRGRKKVVHVLEGESGSVVVQTAPGQVVSHRGGTIVLPCRYHYEAAAHGHDGVRLKWTKVVDPLAFTDVFVALGPQHRAFGSYRGRAELQGDGPGDASLVLRNVTSpecifications
NP_075378 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.29kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BRAL2/KIAA1926 | |
HAPLN4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
404037 | |
HAPLN4 (Human) Recombinant Protein (Q01) | |
QRGRKKVVHVLEGESGSVVVQTAPGQVVSHRGGTIVLPCRYHYEAAAHGHDGVRLKWTKVVDPLAFTDVFVALGPQHRAFGSYRGRAELQGDGPGDASLVLRNVT | |
RUO | |
HAPLN4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |