Learn More
Abnova™ Human GZMM Partial ORF (NP_005308, 85 a.a. - 193 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003004-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Human natural killer (NK) cells and activated lymphocytes express and store a distinct subset of neutral serine proteases together with proteoglycans and other immune effector molecules in large cytoplasmic granules. These serine proteases are collectively termed granzymes and include 4 distinct gene products: granzyme A, granzyme B, granzyme H, and Met-ase, also known as granzyme M. [provided by RefSeq]
Sequence: DSPGLTFHIKAAIQHPRYKPVPALENDLALLQLDGKVKPSRTIRPLALPSKRQVVAAGTRCSMAGWGLTHQGGRLSRVLRELDLQVLDTRMCNNSRFWNGSLSPSMVCLSpecifications
NP_005308 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DSPGLTFHIKAAIQHPRYKPVPALENDLALLQLDGKVKPSRTIRPLALPSKRQVVAAGTRCSMAGWGLTHQGGRLSRVLRELDLQVLDTRMCNNSRFWNGSLSPSMVCL | |
RUO | |
GZMM | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3004 | |
GZMM (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LMET1/MET1 | |
GZMM | |
Recombinant | |
wheat germ expression system |