Learn More
Abnova™ Human GTPBP1 Partial ORF (NP_004277, 1 a.a. - 76 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009567-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is upregulated by interferon-gamma and encodes a protein that is a member of the AGP11/GTPBP1 family of GTP-binding proteins. A structurally similar protein has been found in mouse, where disruption of the gene for that protein had no observable phenotype. [provided by RefSeq]
Sequence: MDEGCGETIYVIGQGSDGTEYGLSEADMEASYATVKSMAEQIEADVILLRERQEAGGRVRDYLVRKRVGDNDFLEVSpecifications
NP_004277 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.1kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDEGCGETIYVIGQGSDGTEYGLSEADMEASYATVKSMAEQIEADVILLRERQEAGGRVRDYLVRKRVGDNDFLEV | |
RUO | |
GTPBP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9567 | |
GTPBP1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GP-1/GP1/HSPC018/MGC20069 | |
GTPBP1 | |
Recombinant | |
wheat germ expression system |