Learn More
Abnova™ Human GTF2IRD1 Full-length ORF (AAH18136, 1 a.a. - 959 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH18136 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9569 |
Molecular Weight (g/mol) | 131.01kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16113852
|
Abnova™
H00009569-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16103852
|
Abnova™
H00009569-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene contains five GTF2I-like repeats and each repeat possesses a potential helix-loop-helix (HLH) motif. It may have the ability to interact with other HLH-proteins and function as a transcription factor or as a positive transcriptional regulator under the control of Retinoblastoma protein. This gene is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by deletion of multiple genes at 7q11.23. Alternative splicing of this gene generates at least 2 transcript variants. [provided by RefSeq]
Sequence: MALLGKRCDVPTNGCGPDRWNSAFTRKDEIITSLVSALDSMCSALSKLNAEVACVAVHDESAFVVSTEKGRMFLNARKELQSDFLRFCRGPPWKDPEAEHPKKVQRGEGGGRSLPRSSLEHGSDVYLLRKMVEEVFDVLYSEALGRASVVPLPYERLLREPGLLAVQGLPEGLAFRRPAEYDPKALMAILEHSHRIRFKLKRPLEDGGRDSKALVELNGVSLIPKGSRDCGLHGQAPKVPPQDLPPTATSSSMASFLYSTALPNHAIRELKQEAPSCPLAPSDLGLSRPMPEPKATGAQDFSDCCGQKPTGPGGPLIQNVHASKRILFSIVHDKSEKWDAFIKETEDINTLRECVQILFNSRYAEALGLDHMVPVPYRKIACDPEAVEIVGIPDKIPFKRPCTYGVPKLKRILEERHSIHFIIKRMFDERIFTGNKFTKDTTKLEPASPPEDTSAEVSRATVLDLAGNARSDKGSMSEDCGPGTSGELGGLRPIKIEPEDLDIIQVTVPDPSPTSEEMTDSMPGHLPSEDSGYGMEMLTDKGLSEDARPEERPVEDSHGDVIRPLRKQVELLFNTRYAKAIGISEPVKVPYSKFLMHPEELFVVGLPEGISLRRPNCFGIAKLRKILEASNSIQFVIKRPELLTEGVKEPIVDSQGTASSLGFSPPALPPERDSGDPLVDESLKRQGFQENYDARLSRIDIANTLREQVQDLFNKKYGEALGIKYPVQVPYKRIKSNPGSVIIEGLPPGIPFRKPCTFGSQNLERILAVADKIKFTVTRPFQGLIPKPDEDDANRLGEKVILREQVKELFNEKYGEALGLNRPVLVPYKLIRDSPDAVEVTGLPDDIPFRNPNTYDIHRLEKILKAREHVRMVIINQLQPFAEICNDAKVPAKDSSIPKRKRKRVSEGNSVSSSSSSSSSSSSNPDSVASANQISFVQWPMYMVDYAGLNVQLPGPLNYSpecifications
AAH18136 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
131.01kDa | |
Glutathione Sepharose 4 Fast Flow | |
MALLGKRCDVPTNGCGPDRWNSAFTRKDEIITSLVSALDSMCSALSKLNAEVACVAVHDESAFVVSTEKGRMFLNARKELQSDFLRFCRGPPWKDPEAEHPKKVQRGEGGGRSLPRSSLEHGSDVYLLRKMVEEVFDVLYSEALGRASVVPLPYERLLREPGLLAVQGLPEGLAFRRPAEYDPKALMAILEHSHRIRFKLKRPLEDGGRDSKALVELNGVSLIPKGSRDCGLHGQAPKVPPQDLPPTATSSSMASFLYSTALPNHAIRELKQEAPSCPLAPSDLGLSRPMPEPKATGAQDFSDCCGQKPTGPGGPLIQNVHASKRILFSIVHDKSEKWDAFIKETEDINTLRECVQILFNSRYAEALGLDHMVPVPYRKIACDPEAVEIVGIPDKIPFKRPCTYGVPKLKRILEERHSIHFIIKRMFDERIFTGNKFTKDTTKLEPASPPEDTSAEVSRATVLDLAGNARSDKGSMSEDCGPGTSGELGGLRPIKIEPEDLDIIQVTVPDPSPTSEEMTDSMPGHLPSEDSGYGMEMLTDKGLSEDARPEERPVEDSHGDVIRPLRKQVELLFNTRYAKAIGISEPVKVPYSKFLMHPEELFVVGLPEGISLRRPNCFGIAKLRKILEASNSIQFVIKRPELLTEGVKEPIVDSQGTASSLGFSPPALPPERDSGDPLVDESLKRQGFQENYDARLSRIDIANTLREQVQDLFNKKYGEALGIKYPVQVPYKRIKSNPGSVIIEGLPPGIPFRKPCTFGSQNLERILAVADKIKFTVTRPFQGLIPKPDEDDANRLGEKVILREQVKELFNEKYGEALGLNRPVLVPYKLIRDSPDAVEVTGLPDDIPFRNPNTYDIHRLEKILKAREHVRMVIINQLQPFAEICNDAKVPAKDSSIPKRKRKRVSEGNSVSSSSSSSSSSSSNPDSVASANQISFVQWPMYMVDYAGLNVQLPGPLNY | |
RUO | |
GTF2IRD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9569 | |
GTF2IRD1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BEN/CREAM1/GTF3/MUSTRD1/RBAP2/WBS/WBSCR11/WBSCR12/hMusTRD1alpha1 | |
GTF2IRD1 | |
Recombinant | |
wheat germ expression system |