missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GSC Partial ORF (NP_776248, 151 a.a. - 257 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00145258-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192007

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

This gene encodes a member of the bicoid subfamily of the paired (PRD) homeobox family of proteins. The encoded protein acts as a transcription factor and may be autoregulatory. A similar protein in mice plays a role in craniofacial and rib cage development during embryogenesis. [provided by RefSeq]

Sequence: LLNQLHCRRKRRHRTIFTDEQLEALENLFQETKYPDVGTREQLARKVHLREEKVEVWFKNRRAKWRRQKRSSSEESENAEKWNKTSSSKASPEKREEEGKSDLDSDS
Specifications

Specifications

NP_776248
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.51kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LLNQLHCRRKRRHRTIFTDEQLEALENLFQETKYPDVGTREQLARKVHLREEKVEVWFKNRRAKWRRQKRSSSEESENAEKWNKTSSSKASPEKREEEGKSDLDSDS
RUO
GSC
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
145258
GSC (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GSC
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers