missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GRID1 Partial ORF (NP_060021, 349 a.a. - 441 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002894-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16125751

  • 335.00€ / 10µg
Estimated Shipment: 06-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a subunit of glutamate receptor channels. These channels mediate most of the fast excitatory synaptic transmission in the central nervous system and play key roles in synaptic plasticity

Sequence: LNCIRKSTKPWNGGRSMLDTIKKGHITGLTGVMEFREDSSNPYVQFEILGTTYSETFGKDMRKLATWDSEKGLNGSLQERPMGSRLQGLTLKV
Specifications

Specifications

NP_060021
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.97kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LNCIRKSTKPWNGGRSMLDTIKKGHITGLTGVMEFREDSSNPYVQFEILGTTYSETFGKDMRKLATWDSEKGLNGSLQERPMGSRLQGLTLKV
RUO
GRID1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
2894
GRID1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA1220
GRID1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers