Learn More
ACROBiosystems Human Gremlin / GREM1 Protein, Fc Tag
Brand: ACROBiosystems GR1-H5254-1MG
Dettagli aggiuntivi : Weight : 0.00001kg
Descrizione
Gremlin is also known as Cysteine knot superfamily 1, BMP antagonist 1 (CKTSF1B1), DAN domain family member 2 (DAND2), Down-regulated in Mos-transformed cells protein (DRM), Increased in high glucose protein 2 (IHG-2), Cell proliferation-inducing gene 2 protein (PIG2) or Gremlin-1 (GREM1), which is highly expressed in small intestine, fetal brain and colon. Gremlin / GREM-1 interacts with SLIT1 and SLIT2 in a glycosylation-dependent manner. Gremlin may play an important role during carcinogenesis and metanephric kidney organogenesis, as a BMP antagonist required for early limb outgrowth and patterning in maintaining the FGF4-SHH feedback loop. Gremlin down-regulates the BMP4 signaling in a dose-dependent manner and acts as inhibitor of monocyte chemotaxis.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 44.2 kDa. The protein migrates as 48-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifica
NP_037504 | |
1 mg | |
Lys 25 - Asp 184 | |
GREM1,CKTSF1B1,DAND2,DRM,PIG2 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
44.2 kDa | |
Human Gremlin, Fc Tag (GR1-H5254) is expressed from human 293 cells (HEK293). It contains AA Lys 25 - Asp 184 (Accession # NP_037504). | |
-20°C | |
Gremlin | |
Human | |
50 mM Tris, 150 mM NaCl, pH7.5 with 1 mM EDTA | |
HEK293 | |
Cytokines and Growth Factors |