missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GPC1 Partial ORF (NP_002072, 24 a.a. - 131 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002817-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16115651

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation. [provided by RefSeq]

Sequence: DPASKSRSCGEVRQIYGAKGFSLSDVPQAEISGEHLRICPQGYTCCTSEMEENLANRSHAELETALRDSSRVLQAMLATQLRSFDDHFQHLLNDSERTLQATFPGAFG
Specifications

Specifications

NP_002072
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.62kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DPASKSRSCGEVRQIYGAKGFSLSDVPQAEISGEHLRICPQGYTCCTSEMEENLANRSHAELETALRDSSRVLQAMLATQLRSFDDHFQHLLNDSERTLQATFPGAFG
RUO
GPC1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
2817
GPC1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ38078/glypican
GPC1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers