missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GPA33 Full-length ORF (NP_005805.1, 1 a.a. - 319 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010223-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16114672

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

The glycoprotein encoded by this gene is a cell surface antigen that is expressed in greater than 95% of human colon cancers. The open reading frame encodes a 319-amino acid polypeptide having a putative secretory signal sequence and 3 potential glycosylation sites. The predicted mature protein has a 213-amino acid extracellular region, a single transmembrane domain, and a 62-amino acid intracellular tail. The sequence of the extracellular region contains 2 domains characteristic of the CD2 subgroup of the immunoglobulin (Ig) superfamily. [provided by RefSeq]

Sequence: MVGKMWPVLWTLCAVRVTVDAISVETPQDVLRASQGKSVTLPCTYHTSTSSREGLIQWDKLLLTHTERVVIWPFSNKNYIHGELYKNRVSISNNAEQSDASITIDQLTMADNGTYECSVSLMSDLEGNTKSRVRLLVLVPPSKPECGIEGETIIGNNIQLTCQSKEGSPTPQYSWKRYNILNQEQPLAQPASGQPVSLKNISTDTSGYYICTSSNEEGTQFCNITVAVRSPSMNVALYVGIAVGVVAALIIIGIIIYCCCCRGKDDNTEDKEDARPNREAYEEPPEQLRELSREREEEDDYRQEEQRSTGRESPDHLDQ
Specifications

Specifications

NP_005805.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
A33/MGC129986/MGC129987
GPA33
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10223
GPA33 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVGKMWPVLWTLCAVRVTVDAISVETPQDVLRASQGKSVTLPCTYHTSTSSREGLIQWDKLLLTHTERVVIWPFSNKNYIHGELYKNRVSISNNAEQSDASITIDQLTMADNGTYECSVSLMSDLEGNTKSRVRLLVLVPPSKPECGIEGETIIGNNIQLTCQSKEGSPTPQYSWKRYNILNQEQPLAQPASGQPVSLKNISTDTSGYYICTSSNEEGTQFCNITVAVRSPSMNVALYVGIAVGVVAALIIIGIIIYCCCCRGKDDNTEDKEDARPNREAYEEPPEQLRELSREREEEDDYRQEEQRSTGRESPDHLDQ
RUO
GPA33
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers