missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GORASP1 Full-length ORF (NP_114105.1, 1 a.a. - 440 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00064689-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193663

  • 528.00€ / 25µg

Estimated Shipment: 30-10-2023
to see stock.
Add to basket

Description

Description

The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a membrane protein involved in establishing the stacked structure of the Golgi apparatus. It is a caspase-3 substrate, and cleavage of this encoded protein contributes to Golgi fragmentation in apoptosis. This encoded protein can form a complex with the Golgi matrix protein GOLGA2, and this complex binds to the vesicle docking protein p115. Several alternatively spliced transcript variants of this gene have been identified, but their full-length natures have not been determined. [provided by RefSeq]

Sequence: MGLGVSAEQPAGGAEGFHLHGVQENSPAQQAGLEPYFDFIITIGHSRLNKENDTLKALLKANVEKPVKLEVFNMKTMRVREVEVVPSNMWGGQGLLGASVRFCSFRRASEQVWHVLDVEPSSPAALAGLRPYTDYVVGSDQILQESEDFFTLIESHEGKPLKLMVYNSKSDSCREVTVTPNAAWGGEGSLGCGIGYGYLHRIPTQPPSYHKKPPGTPPPSALPLGAPPPDALPPGPTPEDSPSLETGSRQSDYMEALLQAPGSSMEDPLPGPGSPSHSAPDPDGLPHFMETPLQPPPPVQRVMDPGFLDVSGISLLDNSNASVWPSLPSSTELTTTAVSTSGPEDICSSSSSHERGGEATWSGSEFEVSFLDSPGAQAQADHLPQLTLPDSLTSAASPEDGLSAELLEAQAEEEPASTEGLDTGTEAEGLDSQAQISTTE
Specifications

Specifications

NP_114105.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
72.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ23443/GOLPH5/GRASP65/MGC118894/MGC118897/P65
GORASP1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
64689
GORASP1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGLGVSAEQPAGGAEGFHLHGVQENSPAQQAGLEPYFDFIITIGHSRLNKENDTLKALLKANVEKPVKLEVFNMKTMRVREVEVVPSNMWGGQGLLGASVRFCSFRRASEQVWHVLDVEPSSPAALAGLRPYTDYVVGSDQILQESEDFFTLIESHEGKPLKLMVYNSKSDSCREVTVTPNAAWGGEGSLGCGIGYGYLHRIPTQPPSYHKKPPGTPPPSALPLGAPPPDALPPGPTPEDSPSLETGSRQSDYMEALLQAPGSSMEDPLPGPGSPSHSAPDPDGLPHFMETPLQPPPPVQRVMDPGFLDVSGISLLDNSNASVWPSLPSSTELTTTAVSTSGPEDICSSSSSHERGGEATWSGSEFEVSFLDSPGAQAQADHLPQLTLPDSLTSAASPEDGLSAELLEAQAEEEPASTEGLDTGTEAEGLDSQAQISTTE
RUO
GORASP1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers