Learn More
Abnova™ Human GOLPH3 Full-length ORF (NP_071413.1, 1 a.a. - 298 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064083-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a peripheral membrane protein of the Golgi stack and may have a regulatory role in Golgi trafficking. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of these variants has not been determined. [provided by RefSeq]
Sequence: MTSLTQRSSGLVQRRTEASRNAADKERAAGGGAGSSEDDAQSRRDEQDDDDKGDSKETRLTLMEEVLLLGLKDREGYTSFWNDCISSGLRGCMLIELALRGRLQLEACGMRRKSLLTRKVICKSDAPTGDVLLDEALKHVKETQPPETVQNWIELLSGETWNPLKLHYQLRNVRERLAKNLVEKGVLTTEKQNFLLFDMTTHPLTNNNIKQRLIKKVQEAVLDKWVNDPHRMDRRLLALIYLAHASDVLENAFAPLLDEQYDLATKRVRQLLDLDPEVECLKANTNEVLWAVVAAFTKSpecifications
NP_071413.1 | |
Liquid | |
64083 | |
GOLPH3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTSLTQRSSGLVQRRTEASRNAADKERAAGGGAGSSEDDAQSRRDEQDDDDKGDSKETRLTLMEEVLLLGLKDREGYTSFWNDCISSGLRGCMLIELALRGRLQLEACGMRRKSLLTRKVICKSDAPTGDVLLDEALKHVKETQPPETVQNWIELLSGETWNPLKLHYQLRNVRERLAKNLVEKGVLTTEKQNFLLFDMTTHPLTNNNIKQRLIKKVQEAVLDKWVNDPHRMDRRLLALIYLAHASDVLENAFAPLLDEQYDLATKRVRQLLDLDPEVECLKANTNEVLWAVVAAFTK | |
RUO | |
GOLPH3 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
60.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ90675/GOPP1/GPP34/MIDAS | |
GOLPH3 | |
Yes | |
wheat germ expression system |