missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GOLGA2 Partial ORF (AAH06381.1, 1 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002801-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16105631

  • 508.00€ / 25µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein has been postulated to play roles in the stacking of Golgi cisternae and in vesicular transport. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of these variants has not been determined. [provided by RefSeq]

Sequence: EQAEARRQILETMQNDRTTISRALSQNRELKEQLAELQSGFVKLTNENMEITSALQSEQHVKRELGKKLGELQEKLSELKETVELKSQEAQSLQQQRDQYLGHLQQYVAAYQQLTSEKEV
Specifications

Specifications

AAH06381.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.94kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EQAEARRQILETMQNDRTTISRALSQNRELKEQLAELQSGFVKLTNENMEITSALQSEQHVKRELGKKLGELQEKLSELKETVELKSQEAQSLQQQRDQYLGHLQQYVAAYQQLTSEKEV
RUO
GOLGA2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
2801
GOLGA2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GM130/MGC20672
GOLGA2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers