missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GNPAT Full-length ORF (NP_055051.1, 1 a.a. - 680 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008443-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16104878

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes an enzyme located in the peroxisomal membrane which is essential to the synthesis of ether phospholipids. Mutations in this gene are associated with rhizomelic chondrodysplasia punctata. [provided by RefSeq]

Sequence: MESSSSSNSYFSVGPTSPSAVVLLYSKELKKWDEFEDILEERRHVSDLKFAMKCYTPLVYKGITPCKPIDIKCSVLNSEEIHYVIKQLSKESLQSVDVLREEVSEILDEMSHKLRLGAIRFCAFTLSKVFKQIFSKVCVNEEGIQKLQRAIQEHPVVLLPSHRSYIDFLMLSFLLYNYDLPVPVIAAGMDFLGMKMVGELLRMSGAFFMRRTFGGNKLYWAVFSEYVKTMLRNGYAPVEFFLEGTRSRSAKTLTPKFGLLNIVMEPFFKREVFDTYLVPISISYDKILEETLYVYELLGVPKPKESTTGLLKARKILSENFGSIHVYFGDPVSLRSLAAGRMSRSSYNLVPRYIPQKQSEDMHAFVTEVAYKMELLQIENMVLSPWTLIVAVLLQNRPSMDFDALVEKTLWLKGLTQAFGGFLIWPDNKPAEEVVPASILLHSNIASLVKDQVILKVDSGDSEVVDGLMLQHITLLMCSAYRNQLLNIFVRPSLVAVALQMTPGFRKEDVYSCFRFLRDVFADEFIFLPGNTLKDFEEGCYLLCKSEAIQVTTKDILVTEKGNTVLEFLVGLFKPFVESYQIICKYLLSEEEDHFSEEQYLAAVRKFTSQLLDQGTSQCYDVLSSDVQKNALAACVRLGVVEKKKINNNCIFNVNEPATTKLEEMLGCKTPIGKPATAKL
Specifications

Specifications

NP_055051.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
103.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DAP-AT/DAPAT/DHAPAT
GNPAT
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
8443
GNPAT (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MESSSSSNSYFSVGPTSPSAVVLLYSKELKKWDEFEDILEERRHVSDLKFAMKCYTPLVYKGITPCKPIDIKCSVLNSEEIHYVIKQLSKESLQSVDVLREEVSEILDEMSHKLRLGAIRFCAFTLSKVFKQIFSKVCVNEEGIQKLQRAIQEHPVVLLPSHRSYIDFLMLSFLLYNYDLPVPVIAAGMDFLGMKMVGELLRMSGAFFMRRTFGGNKLYWAVFSEYVKTMLRNGYAPVEFFLEGTRSRSAKTLTPKFGLLNIVMEPFFKREVFDTYLVPISISYDKILEETLYVYELLGVPKPKESTTGLLKARKILSENFGSIHVYFGDPVSLRSLAAGRMSRSSYNLVPRYIPQKQSEDMHAFVTEVAYKMELLQIENMVLSPWTLIVAVLLQNRPSMDFDALVEKTLWLKGLTQAFGGFLIWPDNKPAEEVVPASILLHSNIASLVKDQVILKVDSGDSEVVDGLMLQHITLLMCSAYRNQLLNIFVRPSLVAVALQMTPGFRKEDVYSCFRFLRDVFADEFIFLPGNTLKDFEEGCYLLCKSEAIQVTTKDILVTEKGNTVLEFLVGLFKPFVESYQIICKYLLSEEEDHFSEEQYLAAVRKFTSQLLDQGTSQCYDVLSSDVQKNALAACVRLGVVEKKKINNNCIFNVNEPATTKLEEMLGCKTPIGKPATAKL
RUO
GNPAT
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers