Learn More
Abnova™ Human GNE Full-length ORF (NP_005467.1, 1 a.a. - 722 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010020-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a bifunctional enzyme that initiates and regulates the biosynthesis of N-acetylneuraminic acid (NeuAc), a precursor of sialic acids. It is a rate-limiting enzyme in the sialic acid biosynthetic pathway. Sialic acid modification of cell surface molecules is crucial for their function in many biologic processes, including cell adhesion and signal transduction. Differential sialylation of cell surface molecules is also implicated in the tumorigenicity and metastatic behavior of malignant cells. Mutations in this gene are associated with sialuria, autosomal recessive inclusion body myopathy, and Nonaka myopathy. Alternative splicing of this gene results in transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MEKNGNNRKLRVCVATCNRADYSKLAPIMFGIKTEPEFFELDVVVLGSHLIDDYGNTYRMIEQDDFDINTRLHTIVRGEDEAAMVESVGLALVKLPDVLNRLKPDIMIVHGDRFDALALATSAALMNIRILHIEGGEVSGTIDDSIRHAITKLAHYHVCCTRSAEQHLISMCEDHDRILLAGCPSYDKLLSAKNKDYMSIIRMWLGDDVKSKDYIVALQHPVTTDIKHSIKMFELTLDALISFNKRTLVLFPNIDAGSKEMVRVMRKKGIEHHPNFRAVKHVPFDQFIQLVAHAGCMIGNSSCGVREVGAFGTPVINLGTRQIGRETGENVLHVRDADTQDKILQALHLQFGKQYPCSKIYGDGNAVPRILKFLKSIDLQEPLQKKFCFPPVKENISQDIDHILETLSALAVDLGGTNLRVAIVSMKGEIVKKYTQFNPKTYEERINLILQMCVEAAAEAVKLNCRILGVGISTGGRVNPREGIVLHSTKLIQEWNSVDLRTPLSDTLHLPVWVDNDGNCAALAERKFGQGKGLENFVTLITGTGIGGGIIHQHELIHGSSFCAAELGHLVVSLDGPDCSCGSHGCIEAYASGMALQREAKKLHDEDLLLVEGMSVPKDEAVGALHLIQAAKLGNAKAQSILRTAGTALGLGVVNILHTMNPSLVILSGVLASHYIHIVKDVIRQQALSSVQDVDVVVSDLVDPALLGAASMVLDYTTRRIYSpecifications
NP_005467.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
105.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DMRV/GLCNE/IBM2/NM/Uae1 | |
GNE | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10020 | |
GNE (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEKNGNNRKLRVCVATCNRADYSKLAPIMFGIKTEPEFFELDVVVLGSHLIDDYGNTYRMIEQDDFDINTRLHTIVRGEDEAAMVESVGLALVKLPDVLNRLKPDIMIVHGDRFDALALATSAALMNIRILHIEGGEVSGTIDDSIRHAITKLAHYHVCCTRSAEQHLISMCEDHDRILLAGCPSYDKLLSAKNKDYMSIIRMWLGDDVKSKDYIVALQHPVTTDIKHSIKMFELTLDALISFNKRTLVLFPNIDAGSKEMVRVMRKKGIEHHPNFRAVKHVPFDQFIQLVAHAGCMIGNSSCGVREVGAFGTPVINLGTRQIGRETGENVLHVRDADTQDKILQALHLQFGKQYPCSKIYGDGNAVPRILKFLKSIDLQEPLQKKFCFPPVKENISQDIDHILETLSALAVDLGGTNLRVAIVSMKGEIVKKYTQFNPKTYEERINLILQMCVEAAAEAVKLNCRILGVGISTGGRVNPREGIVLHSTKLIQEWNSVDLRTPLSDTLHLPVWVDNDGNCAALAERKFGQGKGLENFVTLITGTGIGGGIIHQHELIHGSSFCAAELGHLVVSLDGPDCSCGSHGCIEAYASGMALQREAKKLHDEDLLLVEGMSVPKDEAVGALHLIQAAKLGNAKAQSILRTAGTALGLGVVNILHTMNPSLVILSGVLASHYIHIVKDVIRQQALSSVQDVDVVVSDLVDPALLGAASMVLDYTTRRIY | |
RUO | |
GNE | |
Wheat Germ (in vitro) | |
GST | |
Liquid |