Learn More
Abnova™ Human GNB5 Full-length ORF (NP_006569.1, 1 a.a. - 353 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010681-P02.25ug
Additional Details : Weight : 0.00010kg
Description
Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors. Alternatively spliced transcript variants encoding different isoforms exist. [provided by RefSeq]
Sequence: MATEGLHENETLASLKSEAESLKGKLEEERAKLHDVELHQVAERVEALGQFVMKTRRTLKGHGNKVLCMDWCKDKRRIVSSSQDGKVIVWDSFTTNKEHAVTMPCTWVMACAYAPSGCAIACGGLDNKCSVYPLTFDKNENMAAKKKSVAMHTNYLSACSFTNSDMQILTASGDGTCALWDVESGQLLQSFHGHGADVLCLDLAPSETGNTFVSGGCDKKAMVWDMRSGQCVQAFETHESDINSVRYYPSGDAFASGSDDATCRLYDLRADREVAIYSKESIIFGASSVDFSLSGRLLFAGYNDYTINVWDVLKGSRVSILFGHENRVSTLRVSPDGTAFCSGSWDHTLRVWASpecifications
NP_006569.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ37457/FLJ43714/GB5 | |
GNB5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10681 | |
GNB5 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATEGLHENETLASLKSEAESLKGKLEEERAKLHDVELHQVAERVEALGQFVMKTRRTLKGHGNKVLCMDWCKDKRRIVSSSQDGKVIVWDSFTTNKEHAVTMPCTWVMACAYAPSGCAIACGGLDNKCSVYPLTFDKNENMAAKKKSVAMHTNYLSACSFTNSDMQILTASGDGTCALWDVESGQLLQSFHGHGADVLCLDLAPSETGNTFVSGGCDKKAMVWDMRSGQCVQAFETHESDINSVRYYPSGDAFASGSDDATCRLYDLRADREVAIYSKESIIFGASSVDFSLSGRLLFAGYNDYTINVWDVLKGSRVSILFGHENRVSTLRVSPDGTAFCSGSWDHTLRVWA | |
RUO | |
GNB5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |