missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GNB5 Full-length ORF (AAH11671, 1 a.a. - 126 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010681-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16105572

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors. Alternatively spliced transcript variants encoding different isoforms exist. [provided by RefSeq]

Sequence: MATEGLHENETLASLKSEAESLKGKLEEERAKLHDVELHQVAERVEALGQFVMKTRRTLKGHGNKVLCMDWCKDKRRIVSSSQDGKVIVWDSFTTNKVRHCSQPRYRGFRIPAYLKVLWSSCPALS
Specifications

Specifications

AAH11671
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
39.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ37457/FLJ43714/GB5
GNB5
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10681
GNB5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATEGLHENETLASLKSEAESLKGKLEEERAKLHDVELHQVAERVEALGQFVMKTRRTLKGHGNKVLCMDWCKDKRRIVSSSQDGKVIVWDSFTTNKVRHCSQPRYRGFRIPAYLKVLWSSCPALS
RUO
GNB5
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers