missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human GM-CSF DuoSet ELISA
Click to view available options
Quantity:
5 Plates
Unit Size:
1 set
Description
Includes
Human GM-CSF Capture Antibody; Human GM-CSF Detection Antibody; Human GM-CSF Standard; Streptavidin-HRP
The Human GM-CSF DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human GM-CSF.
Specifications
Specifications
| Conjugate | Biotin |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Quantity | 5 Plates |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction