Learn More
Abnova™ Human GLYAT Full-length ORF (AAH09785.1, 1 a.a. - 163 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010249-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The glycine-N-acyltransferase protein conjugates glycine with acyl-CoA substrates in the mitochondria. The protein is thought to be important in the detoxification of endogenous and xenobiotic acyl-CoA's. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MMLPLQGAQMLQMLEKTLRKSLPASLKVYGTVFHINHGNPFNLKAVVDKWPDFNTVVVCPQEQDMTDDLDHYTNTYQIYSKDPQNCQEFLGSPELINWKQHLQIQSSQPSLNEAIQNLAAIKSFKVKQTQRILYMAAETAKELTPFLLKSKILSPSGGKPKAMSpecifications
AAH09785.1 | |
Liquid | |
10249 | |
GLYAT (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MMLPLQGAQMLQMLEKTLRKSLPASLKVYGTVFHINHGNPFNLKAVVDKWPDFNTVVVCPQEQDMTDDLDHYTNTYQIYSKDPQNCQEFLGSPELINWKQHLQIQSSQPSLNEAIQNLAAIKSFKVKQTQRILYMAAETAKELTPFLLKSKILSPSGGKPKAM | |
RUO | |
GLYAT | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACGNAT/CAT/GAT | |
GLYAT | |
Yes | |
wheat germ expression system |