missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GLTSCR2 Partial ORF (NP_056525, 402 a.a. - 478 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00029997-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: KPRRLGRLKYQAPDIDVQLSSELTDSLRTLKPEGNILRDRFKSFQRRNMIEPRERAKFKRKYKVKLVEKRAFREIQLSpecifications
NP_056525 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.21kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KPRRLGRLKYQAPDIDVQLSSELTDSLRTLKPEGNILRDRFKSFQRRNMIEPRERAKFKRKYKVKLVEKRAFREIQL | |
RUO | |
GLTSCR2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
29997 | |
GLTSCR2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PICT-1/PICT1 | |
GLTSCR2 | |
Recombinant | |
wheat germ expression system |