Learn More
Abnova™ Human GJB1 Partial ORF (AAH22426, 75 a.a. - 174 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH22426 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2705 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16165451
|
Abnova™
H00002705-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16155451
|
Abnova™
H00002705-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]
Sequence: RLWSLQLILVSTPALLVAMHVAHQQHIEKKMLRLEGHGDPLHLEEVKRHKVHISGTLWWTYVISVVFRLLFEAVFMYVFYLLYPGYAMVRLVKCDVYPCPSpecifications
AAH22426 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CMTX/CMTX1/CX32 | |
GJB1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
2705 | |
GJB1 (Human) Recombinant Protein (Q01) | |
RLWSLQLILVSTPALLVAMHVAHQQHIEKKMLRLEGHGDPLHLEEVKRHKVHISGTLWWTYVISVVFRLLFEAVFMYVFYLLYPGYAMVRLVKCDVYPCP | |
RUO | |
GJB1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |