Learn More
Abnova™ Human GIMAP6 Full-length ORF (NP_078987.3, 1 a.a. - 292 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00474344-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, IAN subfamily genes are located in a cluster at 7q36.1. Two transcript variants, one protein-coding and the other probably non-protein-coding, have been found for this gene. [provided by RefSeq]
Sequence: MEEEEYEQIPQENPPEELSQDPVLELSGGLREKEQKTPRRLRLILMGKTGSGKSATGNSILGRDVFESKLSTRPVTKTSQRRSREWAGKELEVIDTPNILSPQVSPEVADAICQAIVLSAPGPHAVLLVTQLGRFTDEDQQVVRRLQEVFGVGVLGHTILVFTRKEDLAGGSLEDYVRETNNQALAWLDVTLARRHCGFNNRAQGEEQEAQLRELMEKVEAIMWENEGDYYSNKAYQYTQQNFRLKELQERQVSQGQGSEDVPGEESWLEGLSQIQKESEEAHRCLLGKADLSpecifications
NP_078987.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686A01175/FLJ22690/IAN6/hIAN2 | |
GIMAP6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
474344 | |
GIMAP6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEEEEYEQIPQENPPEELSQDPVLELSGGLREKEQKTPRRLRLILMGKTGSGKSATGNSILGRDVFESKLSTRPVTKTSQRRSREWAGKELEVIDTPNILSPQVSPEVADAICQAIVLSAPGPHAVLLVTQLGRFTDEDQQVVRRLQEVFGVGVLGHTILVFTRKEDLAGGSLEDYVRETNNQALAWLDVTLARRHCGFNNRAQGEEQEAQLRELMEKVEAIMWENEGDYYSNKAYQYTQQNFRLKELQERQVSQGQGSEDVPGEESWLEGLSQIQKESEEAHRCLLGKADL | |
RUO | |
GIMAP6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |