missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GIMAP6 Full-length ORF (NP_078987.3, 1 a.a. - 292 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00474344-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100754

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, IAN subfamily genes are located in a cluster at 7q36.1. Two transcript variants, one protein-coding and the other probably non-protein-coding, have been found for this gene. [provided by RefSeq]

Sequence: MEEEEYEQIPQENPPEELSQDPVLELSGGLREKEQKTPRRLRLILMGKTGSGKSATGNSILGRDVFESKLSTRPVTKTSQRRSREWAGKELEVIDTPNILSPQVSPEVADAICQAIVLSAPGPHAVLLVTQLGRFTDEDQQVVRRLQEVFGVGVLGHTILVFTRKEDLAGGSLEDYVRETNNQALAWLDVTLARRHCGFNNRAQGEEQEAQLRELMEKVEAIMWENEGDYYSNKAYQYTQQNFRLKELQERQVSQGQGSEDVPGEESWLEGLSQIQKESEEAHRCLLGKADL
Specifications

Specifications

NP_078987.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
59.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686A01175/FLJ22690/IAN6/hIAN2
GIMAP6
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
474344
GIMAP6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEEEEYEQIPQENPPEELSQDPVLELSGGLREKEQKTPRRLRLILMGKTGSGKSATGNSILGRDVFESKLSTRPVTKTSQRRSREWAGKELEVIDTPNILSPQVSPEVADAICQAIVLSAPGPHAVLLVTQLGRFTDEDQQVVRRLQEVFGVGVLGHTILVFTRKEDLAGGSLEDYVRETNNQALAWLDVTLARRHCGFNNRAQGEEQEAQLRELMEKVEAIMWENEGDYYSNKAYQYTQQNFRLKELQERQVSQGQGSEDVPGEESWLEGLSQIQKESEEAHRCLLGKADL
RUO
GIMAP6
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers