missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GIF Full-length ORF (NP_005133.2, 1 a.a. - 417 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002694-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16105451

  • 528.00€ / 25µg

Estimated Shipment: 12-07-2023
to see stock.
Add to basket

Description

Description

This gene is a member of the cobalamin transport protein family. It encodes a glycoprotein secreted by parietal cells of the gastric mucosa and is required for adequate absorption of vitamin B12. Vitamin B12 is necessary for erythrocyte maturation and mutations in this gene may lead to congenital pernicious anemia. [provided by RefSeq]

Sequence: MAWFALYLLSLLWATAGTSTQTQSSCSVPSAQEPLVNGIQVLMENSVTSSAYPNPSILIAMNLAGAYNLKAQKLLTYQLMSSDNNDLTIGQLGLTIMALTSSCRDPGDKVSILQRQMENWAPSSPNAEASAFYGPSLAILALCQKNSEATLPIAVRFAKTLLANSSPFNVDTGAMATLALTCMYNKIPVGSEEGYRSLFGQVLKDIVEKISMKIKDNGIIGDIYSTGLAMQALSVTPEPSKKEWNCKKTTDMILNEIKQGKFHNPMSIAQILPSLKGKTYLDVPQVTCSPDHEVQPTLPSNPGPGPTSASNITVIYTINNQLRGVELLFNETINVSVKSGSVLLVVLEEAQRKNPMFKFETTMTSWGLVVSSINNIAENVNHKTYWQFLSGVTPLNEGVADYIPFNHEHITANFTQY
Specifications

Specifications

NP_005133.2
Liquid
2694
GIF (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAWFALYLLSLLWATAGTSTQTQSSCSVPSAQEPLVNGIQVLMENSVTSSAYPNPSILIAMNLAGAYNLKAQKLLTYQLMSSDNNDLTIGQLGLTIMALTSSCRDPGDKVSILQRQMENWAPSSPNAEASAFYGPSLAILALCQKNSEATLPIAVRFAKTLLANSSPFNVDTGAMATLALTCMYNKIPVGSEEGYRSLFGQVLKDIVEKISMKIKDNGIIGDIYSTGLAMQALSVTPEPSKKEWNCKKTTDMILNEIKQGKFHNPMSIAQILPSLKGKTYLDVPQVTCSPDHEVQPTLPSNPGPGPTSASNITVIYTINNQLRGVELLFNETINVSVKSGSVLLVVLEEAQRKNPMFKFETTMTSWGLVVSSINNIAENVNHKTYWQFLSGVTPLNEGVADYIPFNHEHITANFTQY
RUO
GIF
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
71.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IF/IFMH/INF/TCN3
GIF
Yes
wheat germ expression system
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications
Sonderangebote

Sonderangebote