missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GFI1B Full-length ORF (AAH43371.1, 1 a.a. - 330 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008328-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192362

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

Sequence: MPRSFLVKSKKAHTYHQPRVQEDEPLWPPALTPVPRDQAPSNSPVLSTLFPNQCLDWTNLKREPELEQDQNLARMAPAPEGPIVLSRPQDGDSPLSDSPPFYKPSFSWDTLATTYGHSYRQAPSTMQSAFLEHSVSLYGSPLVPSTEPALDFSLRYSPGMDAYHCVKCNKVFSTPHGLEVHVRRSHSGTRPFACDICGKTFGHAVSLEQHTHVHSQERSFECRMCGKAFKRSSTLSTHLLIHSDTRPYPCQFCGKRFHQKSDMKKHTYIHTGEKPHKCQVCGKAFSQSSNLITHSRKHTGFKPFSCELCTKGFQRKVDLRRHRESQHNLK
Specifications

Specifications

AAH43371.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
63.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GFI1B
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
8328
GFI1B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPRSFLVKSKKAHTYHQPRVQEDEPLWPPALTPVPRDQAPSNSPVLSTLFPNQCLDWTNLKREPELEQDQNLARMAPAPEGPIVLSRPQDGDSPLSDSPPFYKPSFSWDTLATTYGHSYRQAPSTMQSAFLEHSVSLYGSPLVPSTEPALDFSLRYSPGMDAYHCVKCNKVFSTPHGLEVHVRRSHSGTRPFACDICGKTFGHAVSLEQHTHVHSQERSFECRMCGKAFKRSSTLSTHLLIHSDTRPYPCQFCGKRFHQKSDMKKHTYIHTGEKPHKCQVCGKAFSQSSNLITHSRKHTGFKPFSCELCTKGFQRKVDLRRHRESQHNLK
RUO
GFI1B
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers