Learn More
Abnova™ Human GDF10 Partial ORF (NP_004953, 379 a.a. - 478 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002662-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in mice suggest that the protein encoded by this gene plays a role in skeletal morphogenesis. [provided by RefSeq]
Sequence: RYLKVDFADIGWNEWIISPKSFDAYYCAGACEFPMPKIVRPSNHATIQSIVRAVGIIPGIPEPCCVPDKMNSLGVLFLDENRNVVLKVYPNMSVDTCACRSpecifications
NP_004953 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RYLKVDFADIGWNEWIISPKSFDAYYCAGACEFPMPKIVRPSNHATIQSIVRAVGIIPGIPEPCCVPDKMNSLGVLFLDENRNVVLKVYPNMSVDTCACR | |
RUO | |
GDF10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2662 | |
GDF10 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BMP-3b/BMP3B | |
GDF10 | |
Recombinant | |
wheat germ expression system |