Learn More
Abnova™ Human GATA5 Partial ORF (NP_536721, 336 a.a. - 397 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00140628-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a transcription factor that contains two GATA-type zinc fingers. The encoded protein is known to bind to hepatocyte nuclear factor-1alpha (HNF-1alpha), and this interaction is essential for cooperative activation of the intestinal lactase-phlorizin hydrolase promoter. In other organisms, similar proteins may be involved in the establishment of cardiac smooth muscle cell diversity. [provided by RefSeq]
Sequence: PVCPGPSMAPQASGQEDDSLAPGHLEFKFEPEDFAFPSTAPSPQAGLRGALRQEAWCALALASpecifications
NP_536721 | |
Liquid | |
140628 | |
GATA5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
bB379O24.1 | |
GATA5 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.56kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PVCPGPSMAPQASGQEDDSLAPGHLEFKFEPEDFAFPSTAPSPQAGLRGALRQEAWCALALA | |
RUO | |
GATA5 | |
Wheat Germ (in vitro) | |
GST |